Assay Details
| Gene Symbol: Lrp2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026278.3 | NP_080554.1 | 1 - 2 | 201 | 67 | |
| GenBank mRNA | AK007202.1 | - | 1 - 2 | 193 | 67 | |
| AK015594.1 | - | 1 - 2 | 194 | 67 | ||
| BC051150.1 | - | 1 - 2 | 201 | 6772814 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007202.1, BC051150.1, NM_026278.3 transcripts.
Set Membership: |
|

Lrp2bp
Genomic Map