| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500013365.mm.1 |
NM_001164600.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013365.mm.1 |
NM_001164600.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013365.mm.1 |
NM_028364.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013365.mm.1 |
NM_028364.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013365.mm.1 |
NM_133691.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013365.mm.1 |
NM_133691.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013365.mm.1 |
TC1500001762.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500013368.mm.1 |
NM_028364.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013368.mm.1 |
NM_028364.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013368.mm.1 |
NM_133691.5.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013368.mm.1 |
NM_133691.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500013368.mm.1 |
TC1500001762.mm.1 |
MTA TC |