Assay Details
| Gene Symbol: Puf60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028364.2 | NP_082640.2 | 4 - 5 | 390 | 93 | |
| GenBank mRNA | AK146534.1 | - | 4 - 5 | 360 | 93 | |
| AK149632.1 | - | 4 - 5 | 356 | 93 | ||
| AK182141.1 | - | 3 - 4 | 232 | 93 | ||
| AK185614.1 | - | 3 - 4 | 232 | 93 | ||
| AK209860.1 | - | 3 - 4 | 235 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Puf60
Genomic Map