Assay Details
| Gene Symbol: Pdgfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008808.3 | NP_032834.1 | 3 - 4 | 444 | 62 | |
| GenBank mRNA | AK017169.1 | - | 1 - 2 | 29 | 62 | |
| AY324648.1 | - | 3 - 4 | 433 | 62 | ||
| BC003817.1 | - | 3 - 4 | 444 | 3495629 | 62 | |
| M29464.1 | - | 3 - 4 | 324 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pdgfa
Genomic Map