Assay Details
| Gene Symbol: Gnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010312.4 | NP_034442.1 | 2 - 3 | 235 | 79 | |
| GenBank mRNA | AK146941.1 | - | 2 - 3 | 259 | 79 | |
| AK151628.1 | - | 2 - 3 | 225 | 79 | ||
| BC029077.1 | - | 1 - 2 | 75 | 4019292 | 79 | |
| BC059942.1 | - | 2 - 3 | 261 | 6529685 | 79 | |
| BC062178.1 | - | 2 - 3 | 218 | 6330944 | 79 | |
| U34960.1 | - | 1 - 2 | 55 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnb2
Genomic Map