Assay Details
| Gene Symbol: Gtf2ird1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081462.2 | NP_001074931.1 | 17 - 18 | 2134 | 159 | |
| NM_001081463.2 | NP_001074932.1 | 17 - 18 | 2134 | 159 | ||
| NM_001081464.2 | NP_001074933.1 | 17 - 18 | 2134 | 159 | ||
| NM_001081465.1 | NP_001074934.1 | 17 - 18 | 2077 | 159 | ||
| NM_001081466.2 | NP_001074935.1 | 17 - 18 | 2134 | 102 | ||
| NM_001081467.2 | NP_001074936.1 | 17 - 18 | 2134 | 159 | ||
| NM_001081468.2 | NP_001074937.1 | 17 - 18 | 2134 | 159 | ||
| NM_001081469.2 | NP_001074938.1 | 17 - 18 | 2134 | 159 | ||
| NM_001081470.2 | NP_001074939.1 | 17 - 18 | 2134 | 159 | ||
| NM_001244936.1 | NP_001231865.1 | 17 - 18 | 2134 | 159 | ||
| NM_020331.3 | NP_065064.2 | 17 - 18 | 2134 | 159 | ||
| GenBank mRNA | AF247161.1 | - | 17 - 18 | 1994 | 159 | |
| AF257475.1 | - | 17 - 18 | 2110 | 159 | ||
| AF260133.1 | - | 17 - 18 | 2105 | 159 | ||
| AF343348.1 | - | 17 - 18 | 2002 | 159 | ||
| AF343349.1 | - | 17 - 18 | 2008 | 102 | ||
| AF343350.1 | - | 17 - 18 | 2004 | 159 | ||
| AF343351.1 | - | 17 - 18 | 2002 | 159 | ||
| AF497637.1 | - | 17 - 18 | 1994 | 159 | ||
| AF497638.1 | - | 17 - 18 | 1994 | 159 | ||
| AF497639.1 | - | 17 - 18 | 1994 | 159 | ||
| AF497640.1 | - | 17 - 18 | 2002 | 159 | ||
| AF497641.1 | - | 17 - 18 | 2008 | 102 | ||
| AF497642.1 | - | 17 - 18 | 2004 | 159 | ||
| AK078488.1 | - | 17 - 18 | 2002 | 159 | ||
| AK079487.1 | - | 17 - 18 | 2074 | 102 | ||
| AK147196.1 | - | 17 - 18 | 2117 | 159 | ||
| AK184689.1 | - | 1 - 2 | 164 | 159 | ||
| AK184797.1 | - | 1 - 2 | 51 | 159 | ||
| AY030287.1 | - | 17 - 18 | 2105 | 159 | ||
| AY030288.1 | - | 17 - 18 | 2105 | 159 | ||
| AY030289.1 | - | 17 - 18 | 2105 | 159 | ||
| BC050822.1 | - | 17 - 18 | 2129 | 6393328 | 159 | |
| BC057091.1 | - | 17 - 18 | 1956 | 5703300 | 159 | |
| BC058964.1 | - | 17 - 18 | 1956 | 5703300 | 159 | |
| BC083150.1 | - | 17 - 18 | 2036 | 30459984 | 159 | |
More Information
Set Membership: |
|

Gtf2ird1
Genomic Map