Assay Details
| Gene Symbol: Pde6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008806.2 | NP_032832.2 | 2 - 3 | 692 | 97 | |
| GenBank mRNA | AK039397.1 | - | 2 - 3 | 404 | 97 | |
| AK044364.1 | - | 2 - 3 | 635 | 97 | ||
| BC048247.1 | - | 2 - 3 | 671 | 97 | ||
| BC058803.1 | - | 2 - 3 | 639 | 97 | ||
| BC129923.1 | - | 2 - 3 | 622 | 40111882 | 97 | |
| BC129924.1 | - | 2 - 3 | 621 | 97 | ||
| BU505563.1 | - | 2 - 3 | 692 | 97 | ||
| X55968.1 | - | 2 - 3 | 625 | 97 | ||
| X60133.1 | - | 2 - 3 | 632 | 97 | ||
| X87952.1 | - | 2 - 3 | 632 | 97 | ||
More Information
Set Membership: |
|

Pde6b
Genomic Map