Assay Details
| Gene Symbol: Pde6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008806.2 | NP_032832.2 | 7 - 8 | 1132 | 85 | |
| GenBank mRNA | AK039397.1 | - | 7 - 8 | 844 | 85 | |
| AK044364.1 | - | 7 - 8 | 1075 | 85 | ||
| BC048247.1 | - | 7 - 8 | 1111 | 85 | ||
| BC058803.1 | - | 7 - 8 | 1079 | 85 | ||
| BC129923.1 | - | 7 - 8 | 1062 | 40111882 | 85 | |
| BC129924.1 | - | 7 - 8 | 1061 | 85 | ||
| X60133.1 | - | 7 - 8 | 1072 | 85 | ||
| X87952.1 | - | 7 - 8 | 1072 | 85 | ||
More Information
Set Membership: |
|

Pde6b
Genomic Map