Assay Details
| Gene Symbol: Rpap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289569.1 | NP_001276498.1 | 12 - 13 | 1966 | 128 | |
| NM_144911.3 | NP_659160.2 | 12 - 13 | 1952 | 128 | ||
| GenBank mRNA | AK034418.1 | - | 12 - 13 | 1968 | 128 | |
| AK145830.1 | - | 12 - 13 | 1927 | 128 | ||
| BC021895.1 | - | 12 - 13 | 1913 | 3481916 | 128 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BM241364.1 | - | Chr. - - [-] on Build | 128 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpap2
Genomic Map