| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500009133.mm.1 |
NM_001163461.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009133.mm.1 |
NM_001289569.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009133.mm.1 |
NM_001289570.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009133.mm.1 |
NM_144911.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009133.mm.1 |
TC0500001080.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500009143.mm.1 |
NM_001163461.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009143.mm.1 |
NM_001163462.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009143.mm.1 |
NM_001289569.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009143.mm.1 |
NM_001289570.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009143.mm.1 |
NM_144911.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009143.mm.1 |
TC0500001080.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500009132.mm.1 |
NM_001163461.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009132.mm.1 |
NM_001289569.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009132.mm.1 |
NM_001289570.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009132.mm.1 |
NM_144911.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009132.mm.1 |
TC0500001080.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500009142.mm.1 |
NM_001163461.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009142.mm.1 |
NM_001163462.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009142.mm.1 |
NM_001289569.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009142.mm.1 |
NM_001289570.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009142.mm.1 |
NM_144911.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009142.mm.1 |
TC0500001080.mm.1 |
MTA TC |