Assay Details
| Gene Symbol: Nsun5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145414.2 | NP_663389.2 | 7 - 8 | 968 | 75 | |
| GenBank mRNA | AF412029.1 | - | 7 - 8 | 961 | 75 | |
| AK079400.1 | - | 7 - 8 | 969 | 75 | ||
| AK149521.1 | - | 7 - 8 | 950 | 75 | ||
| AK156651.1 | - | 7 - 8 | 963 | 75 | ||
| BC051209.1 | - | 3 - 4 | 400 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nsun5
Genomic Map