Assay Details
| Gene Symbol: Arhgap24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146161.3 | NP_666273.1 | 1 - 2 | 467 | 81 | |
| GenBank mRNA | BC025502.1 | - | 1 - 2 | 316 | 5321985 | 81 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC025502.1, NM_146161.3 transcripts.
Set Membership: |
|

Arhgap24
Genomic Map