Assay Details
| Gene Symbol: Fscn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007984.2 | NP_032010.2 | 2 - 3 | 1084 | 68 | |
| GenBank mRNA | AK083935.1 | - | 2 - 3 | 1085 | 68 | |
| AK167670.1 | - | 2 - 3 | 1084 | 68 | ||
| BC010338.1 | - | 2 - 3 | 1070 | 68 | ||
| BC029281.1 | - | 2 - 3 | 1072 | 68 | ||
| BC037137.1 | - | 2 - 3 | 1081 | 68 | ||
| BC052408.1 | - | 2 - 3 | 1073 | 5716179 | 68 | |
| L33726.1 | - | 2 - 3 | 1068 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fscn1
Genomic Map