Assay Details
| Gene Symbol: Rac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009007.2 | NP_033033.1 | 1 - 2 | 229 | 63 | |
| GenBank mRNA | AK008435.1 | - | 1 - 2 | 230 | 63 | |
| AK009017.1 | - | 1 - 2 | 230 | 63 | ||
| AK011072.2 | - | 1 - 2 | 230 | 63 | ||
| AK034601.1 | - | 1 - 2 | 230 | 63 | ||
| AK047969.1 | - | 1 - 2 | 232 | 63 | ||
| AK076023.1 | - | 1 - 2 | 271 | 63 | ||
| AK088825.1 | - | 1 - 2 | 241 | 63 | ||
| AK166383.1 | - | 1 - 2 | 236 | 63 | ||
| AK199407.1 | - | 1 - 2 | 41 | 63 | ||
| BC003828.1 | - | 1 - 2 | 230 | 3593957 | 63 | |
| BC051053.1 | - | 1 - 2 | 229 | 5038182 | 63 | |
| CX242948.1 | - | 1 - 2 | 266 | 63 | ||
| JN971018.1 | - | 1 - 2 | 33 | 63 | ||
| X57277.1 | - | 1 - 2 | 173 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rac1
Genomic Map