Assay Details
| Gene Symbol: Iqce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028833.3 | NP_083109.2 | 6 - 7 | 628 | 78 | |
| GenBank mRNA | AK006472.2 | - | 5 - 6 | 369 | 78 | |
| AK029333.1 | - | 5 - 6 | 392 | 78 | ||
| BC059223.1 | - | 6 - 7 | 628 | 6417270 | 78 | |
| BC065128.1 | - | 5 - 6 | 380 | 78 | ||
More Information
Set Membership: |
|

Iqce
Genomic Map