Assay Details
| Gene Symbol: Cpsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291248.1 | NP_001278177.1 | 6 - 7 | 721 | 60 | |
| GenBank mRNA | AF033201.1 | - | 6 - 7 | 538 | 60 | |
| EU588394.1 | - | 6 - 7 | 577 | 60 | ||
More Information
Set Membership: |
|

Cpsf4
Genomic Map