Assay Details
| Gene Symbol: Tbx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198052.2 | NP_932169.1 | 3 - 4 | 1636 | 80 | |
| GenBank mRNA | AF429310.1 | - | 2 - 3 | 294 | 80 | |
| AK157247.1 | - | 2 - 3 | 1201 | 80 | ||
| AK172118.1 | - | 2 - 3 | 1203 | 80 | ||
| BC096551.1 | - | 3 - 4 | 1636 | 30547736 | 80 | |
| U15567.1 | - | 1 - 2 | 105 | 80 | ||
| U57328.1 | - | 2 - 3 | 321 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbx3
Genomic Map