Assay Details
| Gene Symbol: Eri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026067.3 | NP_080343.4 | 2 - 3 | 447 | 119 | |
| GenBank mRNA | AK014041.1 | - | 2 - 3 | 448 | 119 | |
| AK014410.1 | - | 2 - 3 | 449 | 119 | ||
| AK144223.1 | - | 2 - 3 | 436 | 119 | ||
| AK165026.1 | - | 2 - 3 | 376 | 119 | ||
| AK172005.1 | - | 2 - 3 | 414 | 119 | ||
| BC046412.1 | - | 2 - 3 | 395 | 5354985 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eri1
Genomic Map