Assay Details
| Gene Symbol: Eif2ak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013557.2 | NP_038585.2 | 833 | 71 | ||
| GenBank mRNA | AK156492.1 | - | 792 | 71 | ||
| AK173160.1 | - | 783 | 71 | |||
| AK215017.1 | - | 2 - 3 | 182 | 71 | ||
| AY033898.1 | - | 818 | 71 | |||
| BC028923.1 | - | 805 | 4164417 | 71 | ||
| BC111035.1 | - | 784 | 5357121 | 71 | ||
More Information
Set Membership: |
|

Eif2ak1
Genomic Map