Assay Details
| Gene Symbol: Mzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290453.1 | NP_001277382.1 | 5 - 6 | 1101 | 91 | |
| GenBank mRNA | AK030410.1 | - | 5 - 6 | 1102 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290453.1 transcripts.
Set Membership: |
|

Mzf1
Genomic Map