Assay Details
| Gene Symbol: Opalin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153520.1 | NP_705740.1 | 1 - 2 | 76 | 66 | |
| GenBank mRNA | AK140031.1 | - | 77 | 66 | ||
| BC060980.1 | - | 1 - 2 | 35 | 6533846 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK140031.1, BC060980.1, NM_153520.1 transcripts.
Set Membership: |
|

Opalin
Genomic Map