Assay Details
| Gene Symbol: Zdhhc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028031.3 | NP_082307.1 | 12 - 13 | 1399 | 76 | |
| GenBank mRNA | AK010382.1 | - | 12 - 13 | 1399 | 76 | |
| AK049640.1 | - | 11 - 12 | 1228 | 76 | ||
| AK145855.1 | - | 12 - 13 | 1319 | 76 | ||
| AK146196.1 | - | 12 - 13 | 1371 | 76 | ||
| BC019493.1 | - | 12 - 13 | 1318 | 76 | ||
| BC046599.1 | - | 12 - 13 | 1318 | 6311721 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc13
Genomic Map