Assay Details
| Gene Symbol: Zdhhc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028031.3 | NP_082307.1 | 14 - 15 | 1580 | 100 | |
| GenBank mRNA | AK010382.1 | - | 14 - 15 | 1580 | 100 | |
| AK049640.1 | - | 13 - 14 | 1409 | 100 | ||
| AK145855.1 | - | 14 - 15 | 1500 | 100 | ||
| AK146196.1 | - | 14 - 15 | 1552 | 100 | ||
| BC019493.1 | - | 14 - 15 | 1499 | 100 | ||
| BC046599.1 | - | 14 - 15 | 1499 | 6311721 | 100 | |
More Information
Set Membership: |
|

Zdhhc13
Genomic Map