Assay Details
| Gene Symbol: Zdhhc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028031.3 | NP_082307.1 | 1 - 2 | 133 | 63 | |
| GenBank mRNA | AK010382.1 | - | 1 - 2 | 133 | 63 | |
| AK145855.1 | - | 1 - 2 | 53 | 63 | ||
| AK146196.1 | - | 1 - 2 | 105 | 63 | ||
| BC019493.1 | - | 1 - 2 | 52 | 63 | ||
| BC046599.1 | - | 1 - 2 | 52 | 6311721 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK010382.1, AK145855.1, AK146196.1, BC046599.1, NM_028031.3 transcripts.
Set Membership: |
|

Zdhhc13
Genomic Map