Assay Details
| Gene Symbol: Tph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136084.2 | NP_001129556.1 | 1 - 2 | 119 | 84 | |
| NM_001276372.1 | NP_001263301.1 | 1 - 2 | 119 | 84 | ||
| GenBank mRNA | AF273257.1 | - | 1 - 2 | 120 | 84 | |
| AK146134.1 | - | 1 - 2 | 119 | 84 | ||
| AK146171.1 | - | 1 - 2 | 119 | 84 | ||
| AK157858.1 | - | 1 - 2 | 85 | 84 | ||
| J04758.1 | - | 1 - 2 | 90 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF273257.1, AK146134.1, AK146171.1, AK157858.1, J04758.1, NM_001136084.2, NM_001276372.1 transcripts.
Set Membership: |
|

Tph1
Genomic Map