Assay Details
| Gene Symbol: Tph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136084.2 | NP_001129556.1 | 9 - 10 | 1161 | 63 | |
| NM_009414.3 | NP_033440.1 | 9 - 10 | 1472 | 63 | ||
| GenBank mRNA | AK146134.1 | - | 9 - 10 | 1161 | 63 | |
| AK146171.1 | - | 9 - 10 | 1161 | 63 | ||
| BC072582.1 | - | 9 - 10 | 1472 | 5718460 | 63 | |
| J04758.1 | - | 9 - 10 | 1132 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tph1
Genomic Map