Assay Details
| Gene Symbol: Gtf2h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291075.1 | NP_001278004.1 | 11 - 12 | 1313 | 121 | |
| NM_008186.4 | NP_032212.3 | 10 - 11 | 1346 | 121 | ||
| GenBank mRNA | AK005065.1 | - | 10 - 11 | 1338 | 121 | |
| BC052837.1 | - | 11 - 12 | 1325 | 30049274 | 121 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gtf2h1
Genomic Map