| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700007885.mm.1 |
NM_008186.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007885.mm.1 |
NM_008186.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007885.mm.1 |
TC0700000879.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007887.mm.1 |
TC0700000879.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007886.mm.1 |
TC0700000879.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007889.mm.1 |
NM_008186.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007889.mm.1 |
NM_008186.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007889.mm.1 |
TC0700000879.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700007890.mm.1 |
NM_008186.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007890.mm.1 |
NM_008186.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700007890.mm.1 |
TC0700000879.mm.1 |
MTA TC |