Assay Details
| Gene Symbol: Gtf2h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291075.1 | NP_001278004.1 | 6 - 7 | 777 | 110 | |
| NM_008186.4 | NP_032212.3 | 5 - 6 | 810 | 110 | ||
| GenBank mRNA | BC052837.1 | - | 6 - 7 | 789 | 30049274 | 110 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gtf2h1
Genomic Map