Assay Details
| Gene Symbol: Gpn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024216.1 | NP_077178.1 | 5 - 6 | 677 | 80 | |
| GenBank mRNA | AK016996.1 | - | 5 - 6 | 608 | 80 | |
| AK080720.1 | - | 5 - 6 | 624 | 80 | ||
| AK087968.1 | - | 5 - 6 | 628 | 80 | ||
| AK146783.1 | - | 5 - 6 | 615 | 80 | ||
| BC003341.1 | - | 5 - 6 | 677 | 3257412 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpn3
Genomic Map