Assay Details
| Gene Symbol: Epo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007942.2 | NP_031968.1 | 4 - 5 | 490 | 59 | |
| GenBank mRNA | BC119265.1 | - | 4 - 5 | 490 | 8734014 | 59 |
| BC119271.1 | - | 4 - 5 | 490 | 8734020 | 59 | |
| BC144883.1 | - | 4 - 5 | 490 | 9053431 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
response to hypoxia
heart morphogenesis
regulation of transcription from RNA polymerase II promoter
apoptotic process
embryo implantation
positive regulation of cell proliferation
negative regulation of calcium ion transport into cytosol
peptidyl-serine phosphorylation
erythrocyte differentiation
negative regulation of myeloid cell apoptotic process
positive regulation of tyrosine phosphorylation of Stat5 protein
hemoglobin biosynthetic process
negative regulation of apoptotic process
erythrocyte maturation
positive regulation of neuron differentiation
positive regulation of DNA replication
positive regulation of protein kinase activity
positive regulation of transcription, DNA-templated
positive regulation of Ras protein signal transduction
cardiac muscle tissue morphogenesis
ventricular cardiac muscle tissue morphogenesis
vasculogenesis involved in coronary vascular morphogenesis
visceral serous pericardium development
cellular hyperosmotic response
negative regulation of intrinsic apoptotic signaling pathway in response to osmotic stress
negative regulation of erythrocyte apoptotic process
negative regulation of cation channel activity
erythropoietin receptor binding
hormone activity
protein kinase activator activity
angiogenesis
response to hypoxia
heart morphogenesis
regulation of transcription from RNA polymerase II promoter
apoptotic process
embryo implantation
positive regulation of cell proliferation
negative regulation of calcium ion transport into cytosol
peptidyl-serine phosphorylation
erythrocyte differentiation
negative regulation of myeloid cell apoptotic process
positive regulation of tyrosine phosphorylation of Stat5 protein
hemoglobin biosynthetic process
negative regulation of apoptotic process
erythrocyte maturation
positive regulation of neuron differentiation
positive regulation of DNA replication
positive regulation of protein kinase activity
positive regulation of transcription, DNA-templated
positive regulation of Ras protein signal transduction
cardiac muscle tissue morphogenesis
ventricular cardiac muscle tissue morphogenesis
vasculogenesis involved in coronary vascular morphogenesis
visceral serous pericardium development
cellular hyperosmotic response
negative regulation of intrinsic apoptotic signaling pathway in response to osmotic stress
negative regulation of erythrocyte apoptotic process
negative regulation of cation channel activity
erythropoietin receptor binding
hormone activity
protein kinase activator activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143659 | BC119265.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143659 | BC119271.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143659 | NM_007942.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143659 | BC144883.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143659 | BC144887.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143659 | BC119265.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143659 | BC119271.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143659 | NM_007942.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143659 | BC144883.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P143659 | BC144887.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 262500 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 17244 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 88830 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150063 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143659 | BC119265.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143659 | BC119271.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143659 | NM_007942.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143659 | BC144883.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143659 | BC144887.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143659 | BC119265.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143659 | BC119271.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143659 | NM_007942.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143659 | BC144883.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P143659 | BC144887.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143659 | BC119265.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143659 | BC119271.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143659 | NM_007942.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143659 | BC144883.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143659 | BC144887.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143659 | BC119265.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143659 | BC119271.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143659 | NM_007942.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143659 | BC144883.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P143659 | BC144887.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P444815 | BC119265.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P444815 | BC119271.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P444815 | NM_007942.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P444815 | BC144883.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P444815 | BC144887.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24125997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24125996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24125980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24125978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24125988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24125992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24125987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24125994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24125976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24125974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24125982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24125995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24125984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24125990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24125979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24125981 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P150063 | - | - |
| Whole Mouse Genome G4122A | A_51_P143659 | BC119265.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143659 | BC119271.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143659 | NM_007942.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143659 | BC144883.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143659 | BC144887.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143659 | BC119265.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143659 | BC119271.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143659 | NM_007942.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143659 | BC144883.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P143659 | BC144887.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P444815 | BC119265.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P444815 | BC119271.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P444815 | NM_007942.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P444815 | BC144883.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P444815 | BC144887.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109290 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115006 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P150063 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04137807 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04137811 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04137812 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04137804 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04137809 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04137818 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04137805 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04137806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24125957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04228543 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126007 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24125998 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24125987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04902874 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126015 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04480154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24125994 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04543788 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24125978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24125976 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04137811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24125979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24125988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126017 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04902877 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04137806 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04137805 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04471946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04137804 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04228546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126000 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126024 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24125995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126001 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10534833 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534834 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534836 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534838 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534837 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534835 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10534832 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500031342.mm.1 | NM_007942.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031342.mm.1 | NM_007942.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031342.mm.1 | TC0500003736.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 99374_at | - | - |
| Murine Genome U74v2 Set | 99374_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 706838 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 940180 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 940180 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000940180 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2742822 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2742822 | - | - |
| mouse-6 v1.1 (Illumina) | scl013856.1_67-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 940180 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl013856.1_67-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 940180 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2742822 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2742822 | - | - |

Epo
Genomic Map