Assay Details
| Gene Symbol: Pnkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290764.1 | NP_001277693.1 | 4 - 5 | 666 | 108 | |
| NM_001290766.1 | NP_001277695.1 | 3 - 4 | 758 | 108 | ||
| NM_021549.3 | NP_067524.2 | 3 - 4 | 758 | 108 | ||
| GenBank mRNA | AK049992.1 | - | 3 - 4 | 561 | 108 | |
| AK159535.1 | - | 3 - 4 | 524 | 108 | ||
| AK159770.1 | - | 3 - 4 | 524 | 108 | ||
| AK160020.1 | - | 3 - 4 | 524 | 108 | ||
| AK160160.1 | - | 4 - 5 | 109 | |||
| BC047269.1 | - | 2 - 3 | 808 | 108 | ||
| BC057659.1 | - | 3 - 4 | 560 | 2865792 | 108 | |
| CK628081.1 | - | 3 - 4 | 570 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pnkp
Genomic Map