Assay Details
| Gene Symbol: Pnkp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290764.1 | NP_001277693.1 | 7 - 8 | 910 | 78 | |
| NM_001290767.1 | NP_001277696.1 | 5 - 6 | 665 | 78 | ||
| NM_021549.3 | NP_067524.2 | 6 - 7 | 1002 | 78 | ||
| GenBank mRNA | AF129451.1 | - | 5 - 6 | 660 | 78 | |
| AK049992.1 | - | 6 - 7 | 805 | 78 | ||
| AK159535.1 | - | 6 - 7 | 768 | 78 | ||
| AK159770.1 | - | 6 - 7 | 768 | 78 | ||
| AK160020.1 | - | 6 - 7 | 768 | 78 | ||
| AK160160.1 | - | 7 - 8 | 907 | 78 | ||
| BC057659.1 | - | 6 - 7 | 804 | 2865792 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pnkp
Genomic Map