Assay Details
| Gene Symbol: Irf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016849.4 | NP_058545.1 | 2 - 3 | 436 | 55 | |
| NR_045611.1 | 2 - 3 | 594 | 55 | |||
| GenBank mRNA | AK013944.1 | - | 2 - 3 | 428 | 55 | |
| AK145977.1 | - | 2 - 3 | 569 | 55 | ||
| AK146378.1 | - | 2 - 3 | 415 | 55 | ||
| AK151750.1 | - | 2 - 3 | 416 | 55 | ||
| AK152689.1 | - | 2 - 3 | 416 | 55 | ||
| BC003233.1 | - | 2 - 3 | 395 | 55 | ||
| BC023244.1 | - | 2 - 3 | 562 | 55 | ||
| BC050882.1 | - | 2 - 3 | 298 | 5690948 | 55 | |
| EU545481.1 | - | 1 - 2 | 167 | 55 | ||
| U75839.1 | - | 2 - 3 | 410 | 55 | ||
| U75840.1 | - | 2 - 3 | 293 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Irf3
Genomic Map