Assay Details
| Gene Symbol: Irf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016849.4 | NP_058545.1 | 5 - 6 | 859 | 71 | |
| NR_045611.1 | 5 - 6 | 1017 | 71 | |||
| GenBank mRNA | AK013944.1 | - | 5 - 6 | 851 | 71 | |
| AK145977.1 | - | 5 - 6 | 992 | 71 | ||
| AK146378.1 | - | 5 - 6 | 838 | 71 | ||
| AK151750.1 | - | 5 - 6 | 839 | 71 | ||
| AK152689.1 | - | 5 - 6 | 839 | 71 | ||
| BC050882.1 | - | 5 - 6 | 721 | 5690948 | 71 | |
| EU545481.1 | - | 4 - 5 | 590 | 71 | ||
| U75839.1 | - | 5 - 6 | 833 | 71 | ||
| U75840.1 | - | 5 - 6 | 716 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Irf3
Genomic Map