Assay Details
| Gene Symbol: Nosip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163684.1 | NP_001157156.1 | 6 - 7 | 649 | 61 | |
| GenBank mRNA | AK007962.1 | - | 6 - 7 | 598 | 61 | |
| DV648837.1 | - | 6 - 7 | 649 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nosip
Genomic Map