Assay Details
| Gene Symbol: Prmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045521.1 | 2 - 3 | 196 | 59 | ||
| GenBank mRNA | BC051953.1 | - | 2 - 3 | 132 | 5692474 | 59 |
| BC062964.1 | - | 2 - 3 | 132 | 5692474 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC051953.1, BC062964.1 transcripts.
Set Membership: |
|

Prmt1
Genomic Map