Assay Details
| Gene Symbol: Flt3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102725.1 | 6 - 7 | 565 | 79 | ||
| GenBank mRNA | BC019801.1 | - | 6 - 7 | 550 | 5132319 | 79 |
More Information
Updated Mapping Information
Set Membership: |
|

Flt3l
Genomic Map