Assay Details
| Gene Symbol: Git2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077359.1 | NP_001070827.1 | 10 - 11 | 942 | 71 | |
| NM_001077360.1 | NP_001070828.1 | 9 - 10 | 938 | 71 | ||
| NM_019834.3 | NP_062808.3 | 9 - 10 | 935 | 71 | ||
| GenBank mRNA | AF148693.1 | - | 9 - 10 | 809 | 71 | |
| AK005365.1 | - | 10 - 11 | 888 | 71 | ||
| AK148029.1 | - | 9 - 10 | 942 | 71 | ||
| AK194737.1 | - | 2 - 3 | 81 | 71 | ||
| AK220323.1 | - | 10 - 11 | 942 | 71 | ||
| BC043062.1 | - | 9 - 10 | 938 | 5705391 | 71 | |
| BC056993.1 | - | 9 - 10 | 938 | 5705391 | 71 | |
More Information
Set Membership: |
|

Git2
Genomic Map