Assay Details
| Gene Symbol: Cabp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013879.2 | NP_038907.1 | 1 - 2 | 116 | 61 | |
| GenBank mRNA | AB079604.1 | - | 1 - 2 | 51 | 61 | |
| AF169152.1 | - | 1 - 2 | 116 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cabp1
Genomic Map