Assay Details
| Gene Symbol: Rnf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016698.2 | NP_057907.2 | 11 - 12 | 2238 | 70 | |
| GenBank mRNA | AK004579.1 | - | 11 - 12 | 2236 | 70 | |
| AK034953.1 | - | 11 - 12 | 2237 | 70 | ||
| AK129106.1 | - | 11 - 12 | 2226 | 70 | ||
| AK146728.1 | - | 11 - 12 | 2238 | 70 | ||
| AK149972.1 | - | 11 - 12 | 2239 | 70 | ||
| AK156502.1 | - | 11 - 12 | 2238 | 70 | ||
| AK165617.1 | - | 11 - 12 | 2238 | 70 | ||
| AK168523.1 | - | 11 - 12 | 2233 | 70 | ||
| AK172065.1 | - | 11 - 12 | 2236 | 70 | ||
| AK194865.1 | - | 2 - 3 | 209 | 70 | ||
| BC010342.1 | - | 11 - 12 | 1947 | 3602969 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf10
Genomic Map