Assay Details
| Gene Symbol: Rnf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016698.2 | NP_057907.2 | 13 - 14 | 2504 | 71 | |
| GenBank mRNA | AB026621.1 | - | 13 - 14 | 2503 | 71 | |
| AK004579.1 | - | 13 - 14 | 2502 | 71 | ||
| AK034953.1 | - | 13 - 14 | 2503 | 71 | ||
| AK129106.1 | - | 13 - 14 | 2492 | 71 | ||
| AK146728.1 | - | 13 - 14 | 2504 | 71 | ||
| AK149972.1 | - | 13 - 14 | 2505 | 71 | ||
| AK156502.1 | - | 13 - 14 | 2504 | 71 | ||
| AK165617.1 | - | 13 - 14 | 2504 | 71 | ||
| AK168523.1 | - | 13 - 14 | 2499 | 71 | ||
| AK172065.1 | - | 13 - 14 | 2502 | 71 | ||
| AK196029.1 | - | 1 - 2 | 116 | 71 | ||
| AK197245.1 | - | 1 - 2 | 116 | 71 | ||
| AK218917.1 | - | 1 - 2 | 116 | 71 | ||
| AK219471.1 | - | 1 - 2 | 116 | 71 | ||
| BC010342.1 | - | 13 - 14 | 2213 | 3602969 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf10
Genomic Map