| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500027547.mm.1 |
NM_016698.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027547.mm.1 |
NM_016698.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027547.mm.1 |
TC0500003093.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500027549.mm.1 |
NM_016698.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027549.mm.1 |
NM_016698.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027549.mm.1 |
TC0500003093.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500027552.mm.1 |
NM_016698.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027552.mm.1 |
NM_016698.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027552.mm.1 |
TC0500003093.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500027555.mm.1 |
NM_016698.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027555.mm.1 |
NM_016698.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027555.mm.1 |
TC0500003093.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500027553.mm.1 |
NM_016698.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027553.mm.1 |
NM_016698.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027553.mm.1 |
TC0500003093.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500027554.mm.1 |
NM_016698.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027554.mm.1 |
NM_016698.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027554.mm.1 |
TC0500003093.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500027550.mm.1 |
NM_016698.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027550.mm.1 |
NM_016698.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027550.mm.1 |
TC0500003093.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500027556.mm.1 |
NM_016698.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027556.mm.1 |
NM_016698.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500027556.mm.1 |
TC0500003093.mm.1 |
MTA TC |