Assay Details
| Gene Symbol: Sppl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029012.2 | NP_083288.2 | 4 - 5 | 382 | 104 | |
| GenBank mRNA | AK014709.1 | - | 4 - 5 | 720 | 104 | |
| BC086646.1 | - | 4 - 5 | 747 | 6817386 | 104 | |
| BU613738.1 | - | 4 - 5 | 495 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sppl3
Genomic Map