Assay Details
| Gene Symbol: Sppl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029012.2 | NP_083288.2 | 6 - 7 | 571 | 58 | |
| GenBank mRNA | AK014709.1 | - | 6 - 7 | 909 | 58 | |
| BC023131.1 | - | 2 - 3 | 186 | 58 | ||
| BC086646.1 | - | 6 - 7 | 936 | 6817386 | 58 | |
| CD806412.1 | - | 2 - 3 | 188 | 58 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sppl3
Genomic Map