Assay Details
| Gene Symbol: Gm36693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052522.1 | - | 1 - 2 | 99 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | calcium homeostasis modulator 1 | Calhm1 | EG546729 | -56.1kb | Sense | Chr. 19 - 47141035 - 47144174 on NCBI Build GRCm38 |
| Downstream | SH3 and PX domains 2A | Sh3pxd2a |
2310014D11Rik, AA589508, AI256723, AI413738, C230050L11, EG329070, Fish, Gm5098, Sh3md1, TSK5, Tks5 |
58.1kb | Sense | Chr. 19 - 47260174 - 47464611 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm36693
Genomic Map