Assay Details
| Gene Symbol: 1700095B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040675.1 | 1 - 2 | 321 | 117 | ||
| NR_040676.1 | 1 - 2 | 321 | 117 | |||
| GenBank mRNA | AK006812.1 | - | 1 - 2 | 305 | 117 | |
| AK007075.1 | - | 1 - 2 | 324 | 117 | ||
| AK015608.1 | - | 1 - 2 | 322 | 117 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 30529 |
Gm30529 | -112.8kb | Sense | Chr. 5 - 112643830 - 112680538 on NCBI Build GRCm38 | |
| Downstream |
adrenergic receptor kinase, beta 2 |
Adrbk2 |
4833444A01Rik, AI851927, AW551196, Adrbk-2, Bark-2, GRK3 |
115.0kb | Anti-sense | Chr. 5 - 112910478 - 113015951 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700095B10Rik
Genomic Map