Assay Details
| Gene Symbol: 1700041B01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018845.1 | - | 2 - 3 | 173 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 36877 |
Gm36877 | -25.7kb | Anti-sense | Chr. 15 - 78638613 - 78645475 on NCBI Build GRCm38 | |
| Downstream | MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase | Mfng | AW546563 | 81.1kb | Anti-sense | Chr. 15 - 78755882 - 78773516 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1700041B01Rik
Genomic Map