Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 30d | Mir30d |
Mirn30d, mir-30d, mmu-mir-30d |
-414.2kb | Anti-sense | Chr. 15 - 68341208 - 68341289 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 41341 |
Gm41341 | 121.6kb | Sense | Chr. 15 - 68878321 - 68883576 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Genomic Map