Assay Details
| Gene Symbol: Gm30123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK044715.1 | - | 2 - 3 | 196 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sortilin-related VPS10 domain containing receptor 1 | Sorcs1 |
Sorcs, mSorCS |
Anti-sense | Chr. 19 - 50143301 - 50679136 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 26629 |
Gm26629 | 345.7kb | Sense | Chr. 19 - 50674015 - 50778657 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm30123
Genomic Map